EMAIL THIS PAGE TO A FRIEND
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine
Fucoxanthin induces apoptosis in human cervical cancer cell line HeLa via PI3K/Akt pathway.
PMID 25113250
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6932
Fucoxanthin, carotenoid antioxidant
C42H58O6
16337
Fucoxanthin, analytical standard
C42H58O6