EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Fast and versatile fabrication of PMMA microchip electrophoretic devices by laser engraving.
PMID 25113407
Abstract
This paper describes the effects of different modes and engraving parameters on the dimensions of microfluidic structures produced in PMMA using laser engraving. The engraving modes included raster and vector, while the explored engraving parameters included power, speed, frequency, resolution, line-width, and number of passes. Under the optimum conditions, the technique was applied to produce channels suitable for CE separations. Taking advantage of the possibility to cut-through the substrates, the laser was also used to define solution reservoirs (buffer, sample, and waste) and a PDMS-based decoupler. The final device was used to perform the analysis of a model mixture of phenolic compounds within 200 s with baseline resolution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
135011
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
C6H6O2
36683
2-Aminophenol, PESTANAL®, analytical standard
C6H7NO
09120
2-Aminophenol, purum, ≥98.0% (HPLC)
C6H7NO
A71301
2-Aminophenol, 99%
C6H7NO
Y0001569
Mesalazine impurity C, European Pharmacopoeia (EP) Reference Standard
C6H7NO
C9510
Pyrocatechol, ≥99%
C6H6O2
C3561
Pyrocatechol, plant cell culture tested, BioReagent, ≥99%, powder
C6H6O2
430749
Pyrocatechol, purified by sublimation, ≥99.5%
C6H6O2
RDD007
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
H2NaO4P
S8282
Sodium phosphate monobasic, BioXtra, ≥99.0%
H2NaO4P
S5011
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
H2NaO4P
S2554
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
H2NaO4P
S6566
Sodium phosphate monobasic, Biotechnology Performance Certified, Cell Culture Tested
H2NaO4P
229903
Sodium phosphate monobasic, 99.999% trace metals basis
H2NaO4P
S0751
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
H2NaO4P
71496
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
H2NaO4P
74092
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
H2NaO4P
609773
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
H2Na16O4P