EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Regulation of the extrinsic apoptotic pathway by microRNA-21 in alcoholic liver injury.
PMID 25118289
Abstract
IL-6/Stat3 is associated with the regulation of transcription of key cellular regulatory genes (microRNAs) during different types of liver injury. This study evaluated the role of IL-6/Stat3 in regulating miRNA and miR-21 in alcoholic liver disease. By microarray, we identified that ethanol feeding significantly up-regulated 0.8% of known microRNAs in mouse liver compared with controls, including miR-21. Similarly, the treatment of normal human hepatocytes (N-Heps) and hepatic stellate cells (HSCs) with ethanol and IL-6 significantly increased miR-21 expression. Overexpression of miR-21 decreased ethanol-induced apoptosis in both N-Heps and HSCs. The expression level of miR-21 was significantly increased after Stat3 activation in N-Heps and HSCs, in support of the concept that the 5'-promoter region of miR-21 is regulated by Stat3. Using real time PCR, we confirmed that miR-21 activation is associated with ethanol-linked Stat3 binding of the miR-21 promoter. A combination of bioinformatics, PCR array, dual-luciferase reporter assay, and Western blot analysis revealed that Fas ligand (TNF superfamily, member 6) (FASLG) and death receptor 5 (DR5) are the direct targets of miR-21. Furthermore, inhibition of miR-21 by specific Vivo-Morpholino and knock-out of IL-6 in ethanol-treated mice also increased the expression of DR5 and FASLG in vivo during alcoholic liver injury. The identification of miR-21 as an important regulator of hepatic cell survival, transformation, and remodeling in vitro, as well as its upstream modulators and downstream targets, will provide insight into the involvement of altered miRNA expression in contributing to alcoholic liver disease progression and testing novel therapeutic approaches for human alcoholic liver diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300708
Anti-STAT3 (Ab-705) antibody produced in rabbit, affinity isolated antibody
SAB4300327
Anti-STAT3 (Ab-727) antibody produced in rabbit, affinity isolated antibody
SAB2500993
Anti-STAT3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102317
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
S5933 Anti-STAT3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104461
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB2104912
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
WH0006774M2
Monoclonal Anti-STAT3 antibody produced in mouse, clone 4D6, purified immunoglobulin, buffered aqueous solution
AMAB90777
Monoclonal Anti-STAT3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0492, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90776
Monoclonal Anti-STAT3 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0490, purified immunoglobulin, buffered aqueous glycerol solution
SAB1404414
Monoclonal Anti-STAT3 antibody produced in mouse, clone 1D11-2A11, purified immunoglobulin, buffered aqueous solution
PLA0255
Rabbit anti-STAT3 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB1406487
Anti-STAT3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution