EMAIL THIS PAGE TO A FRIEND
Journal of medical entomology
Enhanced toxicity of binary mixtures of Bacillus thuringiensis subsp. israelensis and three essential oil major constituents to wild Anopheles sinensis (Diptera: Culicidae) and Aedes albopictus (Diptera: Culicidae).
PMID 25118412
Abstract
An assessment was made of the toxicity of 12 insecticides and three essential oils as well as Bacillus thuringiensis subsp. israelensis (Bti) alone or in combination with the oil major constituents (E)-anethole (AN), (E) -cinnamaldehyde (CA), and eugenol (EU; 1:1 ratio) to third instars of bamboo forest-collected Aedes albopictus (Skuse) and rice paddy field-collected Anopheles sinensis Wiedemann. An. sinensis larvae were resistant to various groups of the tested insecticides. Based on 24-h LC50 values, binary mixtures of Bti and CA, AN, or EU were significantly more toxic against Ae. albopictus larvae (0.0084, 0.0134, and 0.0237 mg/liter) and An. sinensis larvae (0.0159, 0.0388, and 0.0541 mg/liter) than either Bti (1.7884 and 2.1681 mg/liter) or CA (11.46 and 18.56 mg/liter), AN (16.66 and 25.11 mg/liter), or EU (24.60 and 31.09 mg/liter) alone. As judged by cotoxicity coefficient (CC) and synergistic factor (SF), the three binary mixtures operated in a synergy pattern (CC, 140.7-368.3 and SF, 0.0007-0.0010 for Ae. albopictus; CC, 75.1-245.3 and SF, 0.0008-0.0017 for An. sinensis). Global efforts to reduce the level of highly toxic synthetic insecticides in the aquatic environment justify further studies on the binary mixtures of Bti and essential oil constituents described, in particular CA, as potential larvicides for the control of malaria vector mosquito populations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W208620
Anethol, natural, 99%, FG
C10H12O
34314
Bifenthrin, PESTANAL®, analytical standard, mixture of isomers
C23H22ClF3O2
37913
Chlorfenapyr, PESTANAL®, analytical standard
C15H11BrClF3N2O
45395
Chlorpyrifos, PESTANAL®, analytical standard
C9H11Cl3NO3PS
94114
Chlorpyrifos, certified reference material, TraceCERT®
C9H11Cl3NO3PS
45423
Deltamethrin, PESTANAL®, analytical standard
C22H19Br2NO3
05995
Deltamethrin, certified reference material, TraceCERT®
C22H19Br2NO3
34094
Etofenprox, PESTANAL®, analytical standard
C25H28O3
79891
Eugenol, certified reference material, TraceCERT®
C10H12O2
01050595
Eugenol, primary reference standard
C10H12O2
E5000000
Eugenol, European Pharmacopoeia (EP) Reference Standard
C10H12O2
45487
Fenitrothion, PESTANAL®, analytical standard
C9H12NO5PS
36552
Fenthion, PESTANAL®, analytical standard
C10H15O3PS2
45614
Permethrin, PESTANAL®, analytical standard
C21H20Cl2O3
442748
Permethrin (isomers), PESTANAL®, analytical standard
C21H20Cl2O3
31526
Temephos, PESTANAL®, analytical standard
C16H20O6P2S3
117870
trans-Anethole, 99%
C10H12O
W208604
trans-Anethole, ≥99%, FCC, FG
C10H12O
PHR1218
trans-Anethole, Pharmaceutical Secondary Standard; Certified Reference Material
C10H12O
10368
trans-Anethole, analytical standard
C10H12O
10370
trans-Anethole, purum, ≥98.0% (GC)
C10H12O