EMAIL THIS PAGE TO A FRIEND
Dentistry today
Endodontic canal preparation: innovations in glide path management and shaping canals.
PMID 25118526
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357553
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Ni
267007
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Ni
268259
Nickel, foil, thickness 0.125 mm, ≥99.9%
Ni
357588
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Ni
203904
Nickel, powder, <150 μm, 99.99% trace metals basis
Ni
266981
Nickel, powder, <50 μm, 99.7% trace metals basis
Ni
268283
Nickel, powder, <1 μm, 99.8% trace metals basis
Ni
577995
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Ni
267074
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Ni
267058
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Ni
357626
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ni
357634
Nickel, wire, diam. 0.25 mm, ≥99.9%
Ni
767484
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ni
GF82156912
Nickel, rod, 100mm, diameter 12.0mm, 99%
Ni
GF11598194
Nickel, rod, 100mm, diameter 12.7mm, 99.98%
Ni
GF29576010
Nickel, rod, 100mm, diameter 16mm, 99.99+%
Ni
GF83584143
Nickel, rod, 100mm, diameter 2.0mm, 99.99+%
Ni
GF45193697
Nickel, rod, 100mm, diameter 2.0mm, annealed, 99%
Ni
GF11517948
Nickel, rod, 100mm, diameter 20.0mm, 99%
Ni
GF39205931
Nickel, rod, 100mm, diameter 25.4mm, 99.98%
Ni
GF69167920
Nickel, rod, 100mm, diameter 3.0mm, 99%
Ni
GF70332870
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Ni
GF99799968
Nickel, rod, 100mm, diameter 30mm, 99%
Ni
GF90754241
Nickel, rod, 100mm, diameter 40mm, 99%
Ni
GF20561385
Nickel, rod, 100mm, diameter 5.0mm, 99.99+%
Ni
GF90932823
Nickel, rod, 100mm, diameter 6.35mm, 99%
Ni
GF50593002
Nickel, rod, 100mm, diameter 9.5mm, 99%
Ni
GF68355085
Nickel, rod, 200mm, diameter 1.5mm, 99.99+%
Ni
GF66250256
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Ni
GF69613825
Nickel, rod, 200mm, diameter 12.0mm, 99%
Ni