EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Relating surface-enhanced Raman scattering signals of cells to gold nanoparticle aggregation as determined by LA-ICP-MS micromapping.
PMID 25120183