EMAIL THIS PAGE TO A FRIEND
International Journal of Clinical and Experimental Pathology
Pathogenesis of fulminant monkeypox with bacterial sepsis after experimental infection with West African monkeypox virus in a cynomolgus monkey.
PMID 25120821