EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Antibody-mediated immune suppression of erythrocyte alloimmunization can occur independently from red cell clearance or epitope masking in a murine model.
PMID 25122924
Abstract
Anti-D can prevent immunization to the RhD Ag on RBCs, a phenomenon commonly termed Ab-mediated immune suppression (AMIS). The most accepted theory to explain this effect has been the rapid clearance of RBCs. In mouse models using SRBC, these xenogeneic cells are always rapidly cleared even without Ab, and involvement of epitope masking of the SRBC Ags by the AMIS-inducing Ab (anti-SRBC) has been suggested. To address these hypotheses, we immunized mice with murine transgenic RBCs expressing the HOD Ag (hen egg lysozyme [HEL], in sequence with ovalbumin, and the human Duffy transmembrane protein) in the presence of polyclonal Abs or mAbs to the HOD molecule. The isotype, specificity, and ability to induce AMIS of these Abs were compared with accelerated clearance as well as steric hindrance of the HOD Ag. Mice made IgM and IgG reactive with the HEL portion of the molecule only. All six of the mAbs could inhibit the response. The HEL-specific Abs (4B7, IgG1; GD7, IgG2b; 2F4, IgG1) did not accelerate clearance of the HOD-RBCs and displayed partial epitope masking. The Duffy-specific Abs (MIMA 29, IgG2a; CBC-512, IgG1; K6, IgG1) all caused rapid clearance of HOD RBCs without steric hindrance. To our knowledge, this is the first demonstration of AMIS to erythrocytes in an all-murine model and shows that AMIS can occur in the absence of RBC clearance or epitope masking. The AMIS effect was also independent of IgG isotype and epitope specificity of the AMIS-inducing Ab.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0529
Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B7151
Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701031 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
B7401
Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B9904
Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B7022 Anti-Mouse IgG (γ-chain specific)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4600004 Anti-Mouse IgG (H+L), Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB3701135 Anti-Mouse IgG (H+L), F(ab′)2 fragment-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701160 Anti-Mouse IgG (H+L), F(ab) fragment-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600005 Anti-Mouse IgG (H+L), highly cross-adsorbed, Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody
B7264
Anti-Mouse IgG (whole molecule)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B6649 Anti-Mouse IgG (whole molecule)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701017 Anti-Mouse IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700995 Anti-Mouse IgG F(ab′)2-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701207 Anti-Mouse IgM (μ-chain specific), F(ab′)2 fragment-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A9688 Anti-Mouse IgM (μ-chain specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
C4431 Citrate-phosphate-dextrose solution with adenine
D8885
Diethanolamine, reagent grade, ≥98.0%
C4H11NO2
31590
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
C4H11NO2
31589
Diethanolamine, BioUltra, ≥99.5% (GC)
C4H11NO2
398179
Diethanolamine, ACS reagent, ≥98.5%
C4H11NO2
16957
Diethanolamine, analytical standard
C4H11NO2
PHR1220
Diethanolamine, Pharmaceutical Secondary Standard; Certified Reference Material
C4H11NO2
Y0001183
Trolamine impurity B, European Pharmacopoeia (EP) Reference Standard
C4H11NO2