EMAIL THIS PAGE TO A FRIEND
Journal of Separation Science
Rapid separation and characterization of diterpenoid alkaloids in processed roots of Aconitum carmichaeli using ultra high performance liquid chromatography coupled with hybrid linear ion trap-Orbitrap tandem mass spectrometry.
PMID 25124198