EMAIL THIS PAGE TO A FRIEND
Journal of Controlled Release : Official Journal of the Controlled Release Society
Probing the size limit for nanomedicine penetration into Burkholderia multivorans and Pseudomonas aeruginosa biofilms.
PMID 25125326