EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
The cholinergic neuronal differentiation factor from heart cells is identical to leukemia inhibitory factor.
PMID 2512641
Abstract
A protein secreted by cultured rat heart cells can direct the choice of neurotransmitter phenotype made by cultured rat sympathetic neurons. Structural analysis and biological assays demonstrated that this protein is identical to a protein that regulates the growth and differentiation of embryonic stem cells and myeloid cells, and that stimulates bone remodeling and acute-phase protein synthesis in hepatocytes. This protein has been termed D factor, DIA, DIF, DRF, HSFIII, and LIF. Thus, this cytokine, like IL-6 and TGF beta, regulates growth and differentiation in the embryo and in the adult in many tissues, now including the nervous system.