EMAIL THIS PAGE TO A FRIEND
The international journal of esthetic dentistry
Comparison of marginal fit of Lava CAD/CAM crown-copings with two finish lines.
PMID 25126621
Abstract
Marginal fit is valued as one of the most important criteria for the clinical quality and success of all-ceramic crowns. The aim of this in vitro study was to investigate the marginal fit of Lava Zirconia crown-copings on chamfer and shoulder preparations. Two acrylic model teeth were selected to simulate the clinical preparations: one molar was prepared with a chamfer finish line (C) and one premolar was prepared with a rounded shoulder finish line (RS). Each resin model was duplicated 10 times using silicon-based impression material and poured in type IV dental stone for the fabrication of working dies. A total of 20 copings were divided into two groups (n = 10 for each finish line). Fifty measuring locations were chosen randomly along the margin on the dies and the gap width - vertical marginal discrepancy - was measured under a light microscope with a magnification of x100. Measurements were made without cementation. The mean marginal gap widths and standard deviations were calculated and a one-way analysis of variance (ANOVA) was performed for different types of preparations in order to detect differences (α = 0.05). The mean marginal gap was 30 ± 3 μm for the C group and 28 ± 4 μm for the RS group. The one-way ANOVA showed no statistical significant difference between the two groups (P = 0.23). Within the limitations of this study, the marginal discrepancies were all within the clinical acceptable standard set at 120 μm. Chamfer and shoulder preparations did not show differences regarding the gap dimension. Bur design is an easily selected parameter before natural tooth preparation. Both tested finish lines are able to help clinicians in obtaining acceptable marginal fit values for the investigated zirconia copings.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
403296
Zirconium, powder, −100 mesh
Zr
267651
Zirconium, sponge, ≥99% trace metals basis
Zr
267724
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Zr
GF54823642
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF32349685
Zirconium, foil, light tested, 25x25mm, thickness 0.025mm, annealed, 99.2%
Zr
GF01998322
Zirconium, foil, light tested, 25x25mm, thickness 0.01mm, 99.2%
Zr
GF56264597
Zirconium, foil, not light tested, 50x50mm, thickness 0.006mm, 99.2%
Zr
GF47333718
Zirconium, rod, 1000mm, diameter 1.5mm, 99.2%
Zr
GF56980165
Zirconium, rod, 1000mm, diameter 12.7mm, centerless ground, 99.2%
Zr
GF67908583
Zirconium, rod, 1000mm, diameter 2.0mm, centerless ground, 99.2%
Zr
GF48459774
Zirconium, rod, 1000mm, diameter 3.0mm, centerless ground, 99.2%
Zr
GF36220227
Zirconium, rod, 1000mm, diameter 5.0mm, centerless ground, 99.2%
Zr
GF70124442
Zirconium, rod, 1000mm, diameter 6.0mm, centerless ground, 99.2%
Zr
GF11484468
Zirconium, rod, 1000mm, diameter 9.5mm, centerless ground, 99.2%
Zr
GF66272081
Zirconium, rod, 100mm, diameter 1.5mm, 99.2%
Zr
GF35558321
Zirconium, rod, 100mm, diameter 12.7mm, centerless ground, 99.2%
Zr
GF15031662
Zirconium, rod, 100mm, diameter 19.0mm, centerless ground, 99.2%
Zr
GF63994708
Zirconium, rod, 100mm, diameter 2.0mm, centerless ground, 99.2%
Zr
GF90246056
Zirconium, rod, 100mm, diameter 3.0mm, centerless ground, 99.2%
Zr
GF88573618
Zirconium, rod, 100mm, diameter 5.0mm, centerless ground, 99.2%
Zr
GF04513700
Zirconium, rod, 100mm, diameter 50mm, centerless ground, 99.2%
Zr
GF27595637
Zirconium, rod, 100mm, diameter 6.0mm, centerless ground, 99.2%
Zr
GF27528048
Zirconium, rod, 100mm, diameter 9.5mm, centerless ground, 99.2%
Zr
GF84010496
Zirconium, rod, 200mm, diameter 1.5mm, 99.2%
Zr
GF44790846
Zirconium, rod, 200mm, diameter 12.7mm, centerless ground, 99.2%
Zr
GF78958715
Zirconium, rod, 200mm, diameter 19.0mm, centerless ground, 99.2%
Zr
GF99338743
Zirconium, rod, 200mm, diameter 2.0mm, centerless ground, 99.2%
Zr
GF92588967
Zirconium, rod, 200mm, diameter 3.0mm, centerless ground, 99.2%
Zr
GF71701512
Zirconium, rod, 200mm, diameter 5.0mm, centerless ground, 99.2%
Zr
GF17709341
Zirconium, rod, 200mm, diameter 6.0mm, centerless ground, 99.2%
Zr