EMAIL THIS PAGE TO A FRIEND
International journal of food microbiology
Development of a selective agar plate for the detection of Campylobacter spp. in fresh produce.
PMID 25126968
Abstract
This study was conducted to develop a selective medium for the detection of Campylobacter spp. in fresh produce. Campylobacter spp. (n=4), non-Campylobacter (showing positive results on Campylobacter selective agar) strains (n=49) isolated from fresh produce, indicator bacteria (n=13), and spoilage bacteria isolated from fresh produce (n=15) were plated on four Campylobacter selective media. Bolton agar and modified charcoal cefoperazone deoxycholate agar (mCCDA) exhibited higher sensitivity for Campylobacter spp. than did Preston agar and Hunt agar, although certain non-Campylobacter strains isolated from fresh produce by using a selective agar isolation method, were still able to grow on Bolton agar and mCCDA. To inhibit the growth of non-Campylobacter strains, Bolton agar and mCCDA were supplemented with 5 antibiotics (rifampicin, polymyxin B, sodium metabisulfite, sodium pyruvate, ferrous sulfate) and the growth of Campylobacter spp. (n=7) and non-Campylobacter strains (n=44) was evaluated. Although Bolton agar supplemented with rifampicin (BR agar) exhibited a higher selectivity for Campylobacter spp. than did mCCDA supplemented with antibiotics, certain non-Campylobacter strains were still able to grow on BR agar (18.8%). When BR agar with various concentrations of sulfamethoxazole-trimethoprim were tested with Campylobacter spp. (n=8) and non-Campylobacter (n=7), sulfamethoxazole-trimethoprim was inhibitory against 3 of 7 non-Campylobacter strains. Finally, we validated the use of BR agar containing 50mg/L sulfamethoxazole (BRS agar) or 0.5mg/L ciprofloxacin (BRCS agar) and other selective agars for the detection of Campylobacter spp. in chicken and fresh produce. All chicken samples were positive for Campylobacter spp. when tested on mCCDA, BR agar, and BRS agar. In fresh produce samples, BRS agar exhibited the highest selectivity for Campylobacter spp., demonstrating its suitability for the detection of Campylobacter spp. in fresh produce.