EMAIL THIS PAGE TO A FRIEND
Talanta
Development, optimization and validation of a multimethod for the determination of 36 mycotoxins in wines by liquid chromatography-tandem mass spectrometry.
PMID 25127606