EMAIL THIS PAGE TO A FRIEND
Chemosphere
Assessment of endosulfan induced genotoxicity and mutagenicity manifested by oxidative stress pathways in freshwater cyprinid fish crucian carp (Carassius carassius L.).
PMID 25128633