EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Identification and quantification of antitumor thioproline and methylthioproline in Korean traditional foods by a liquid chromatography-atmospheric pressure chemical ionization-tandem mass spectrometry.
PMID 25128876
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30095
D-Cysteine, ≥99% (RT)
C3H7NO2S