EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Elevated levels of Th17 cells and Th17-related cytokines are associated with disease activity in patients with inflammatory bowel disease.
PMID 25129403
Abstract
Interleukin-17(IL-17)-producing T helper(Th)17 cells are considered as a new subset of cells critical to the development of inflammatory bowel disease (IBD). We aimed to investigate the distribution of Th17 cells, the expressions of Th17-related cytokines (IL-17, IL-21 and IL-22) and their association with disease activity in IBD patients. We collected intestinal tissue biopsies from 40 patients with active ulcerative colitis (UC), 20 patients with active Crohn's disease (CD) and 20 healthy controls. The distribution of Th17 cells and expressions of Th17-related cytokines in colonic tissues were evaluated by a standard immunohistochemical procedure. Serum IL-17, IL-21 and IL-22 levels were determined by ELISA. Pearson's and Spearman's correlation analyses were performed to analyze the correlation between the number of Th17 cells, the expressions of Th17-related cytokines and disease activity index, endoscopic and histological grading, and CRP and PLT levels, respectively. Compared with healthy controls, the number of Th17 cells and the expressions of IL-17, IL-21 and IL-22 were significantly increased in active IBD patients (P < 0.05). In addition, Pearson's and Spearman's correlation analyses showed that the number of Th17 cells and the expressions of Th17-related cytokines were correlated with disease activity index, endoscopic and histological grading, CRP and PLT levels (P < 0.05). Th17 cells and Th17-related cytokines (IL-17, IL-21 and IL-22) were increased in the intestinal mucosa in active IBD patients and may play an important role in disease activity and mucosal damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS4877
Anti-IL-17 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4887
Anti-IL-17 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500345
Anti-IL-21 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2502114
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502115
Anti-IL17A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1407827
Anti-IL21 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411185
Anti-IL21 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV49523
Anti-IL22 antibody produced in rabbit, affinity isolated antibody
SAB1410873
Anti-IL22 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW21034 Anti-Interleukin-21 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3701439
Monoclonal Anti-IL-17A antibody produced in mouse, clone 4H1524.1, purified immunoglobulin
GW22812 Anti-Interleukin-21 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3701458
Monoclonal Anti-IL-17A antibody produced in rat, clone 20B4.G10.F5, purified immunoglobulin
SAB3501070
Anti-IL-22 antibody produced in rabbit, affinity isolated antibody