EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Ionic liquid intercalated clay sorbents for micro solid phase extraction of steroid hormones from water samples with analysis by liquid chromatography-tandem mass spectrometry.
PMID 25130089
Abstract
Clay material plays an important role in the transport and retention of many compounds in the soil, therefore, clay based sorbents are promising alternatives for selective sorption of organic pollutants. In the present work, different chain length ionic liquids (ILs) namely, 1-methyl-3-octyl-imidazolium bromide, 1-methyl-3-undecyl-imidazolium bromide and 1-methyl-3-octadecyl-imidazolium bromide were intercalated in the galleries of montmorillonite (MMT) clay. Then, this novel nanofiller surface was utilized in micro extraction of estrogenic hormones for the first time. A fast procedure where sonication-assisted emulsification microextraction combined with vortex assisted micro-solid phase extraction (μ-SPE) was developed for the LC-MS/MS analysis of estrone (E1), 17β-estradiol (E2), estriol (E3) and ethynylestradiol (EE2). The parameters related to the μ-SPE procedure namely; pH, sorbent amount, extraction solvent type and volume, sonication and vortex time, sample volume and salt effect on the extraction efficiency were screened by applying Plackett-Burmann design. The selected parameters were then optimized by using Box-Behnken design. The method was validated for the determination of estrogenic hormone residues in river water samples. Linear calibration plots were obtained for all hormones whose regression coefficients were larger than 0.98. RSD values were found less than 10% for three levels of concentration. LOD levels were calculated as; 0.012, 0.062, 0.018 and 0.693 ng L(-1) for E1, E2, E3 and EE2, respectively. Recovery values were calculated in the range of 86.9-97.7%. Considering large sample volumes required for attaining low limits of these hormones, present method provides an ease for analyst as 10 mL of the sample is adequate for achieving the same sensitivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31573
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
C19H30O2
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
294993
Ammonia, anhydrous, ≥99.98%
H3N
779423
Ammonia solution, 4 M in methanol
H3N
718939
Ammonia solution, 0.4 M in THF
H3N
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
227048
Cyclohexane, anhydrous, 99.5%
C6H12
650455
Cyclohexane, for HPLC, ≥99.9%
C6H12
179191
Cyclohexane, ACS reagent, ≥99%
C6H12
34855
Cyclohexane, for HPLC, ≥99.7%
C6H12
33117
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
C6H12
C100307
Cyclohexane, Laboratory Reagent, ≥99.8%
C6H12
676861
Cyclohexane, ACS reagent, ≥99%
C6H12
270989
Ethyl acetate, anhydrous, 99.8%
C4H8O2
58958
Ethyl acetate, analytical standard
C4H8O2
W241415
Ethyl acetate, natural, ≥99%, FCC, FG
C4H8O2
W241407
Ethyl acetate, ≥99%, FCC, FG
C4H8O2
437549
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
537446
Ethyl acetate, ReagentPlus®, ≥99.8%
C4H8O2
270520
Ethyl acetate, HPLC grade, 99.8%
C4H8O2
L092004
Ethyl acetate
C4H8O2
L092003
Ethyl acetate
C4H8O2
PHR1481
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
C4H8O2
1005706
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
C2H4O2
296090
Hexane, anhydrous, 95%
C6H14
52750
Hexane, analytical standard
C6H14
675393
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
C6H14
439177
Hexane, for HPLC, ≥95%
C6H14
248878
Hexane, spectrophotometric grade, ≥95%
C6H14