EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Parallel dual secondary column-dual detection: a further way of enhancing the informative potential of two-dimensional comprehensive gas chromatography.
PMID 25130094
Abstract
Comprehensive two-dimensional gas chromatography (GC×GC) coupled with Mass Spectrometry (MS) is one of today's most powerful analytical platforms for detailed analysis of medium-to-high complexity samples. The column set usually consists of a long, conventional-inner-diameter first dimension ((1)D) (typically 15-30m long, 0.32-0.25mm dc), and a short, narrow-bore second dimension ((2)D) column (typically 0.5-2m, 0.1mm dc) where separation is run in a few seconds. However, when thermal modulation is used, since the columns of a set are coupled in series, a flow mismatch occurs between the two dimensions, making it impossible to operate simultaneously at optimized flow conditions. Further, short narrow-bore capillaries can easily be overloaded, because of their lower loadability, limiting the effectiveness of (2)D separation. In this study, improved gas linear velocities in both chromatographic dimensions were achieved by coupling the (1)D column with two parallel (2)D columns, having identical inner diameter, stationary phase chemistry, and film thickness. In turn, these were connected to two detectors: a fast quadrupole Mass Spectrometer (MS) and a Flame Ionization Detector (FID). Different configurations were tested and performances compared to a conventional set-up; experimental results on two model mixtures (n-alkanes and fourteen medium-to-high polarity volatiles of interest in the flavor and fragrance field) and on the essential oil of Artemisia umbelliformis Lam., show the system provides consistent results, in terms of analyte identification (reliability of spectra and MS matching) and quantitation, also affording an internal cross-validation of quantitation accuracy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21310
(±)-Camphor, purum, synthetic, ≥95.0% (GC)
C10H16O
20506
(±)-Camphor, meets analytical specification of Ph. Eur., BP, racemic, ≥95% (GC)
C10H16O
W526606
(±)-Camphor, ≥95.5%
C10H16O
21300
(+)-Camphor, purum, natural, ≥97.0% (sum of enantiomers, GC)
C10H16O
21293
(−)-Camphor, analytical standard
C10H16O
21295
(−)-Camphor, purum, ≥95.0% (sum of enantiomers, GC)
C10H16O
418099
Benzaldehyde, purified by redistillation, ≥99.5%
C7H6O
B1334
Benzaldehyde, ReagentPlus®, ≥99%
C7H6O
09143
Benzaldehyde, analytical standard
C7H6O
PHR1203
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O
Y0001064
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
C7H6O
12010
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
C7H6O
W212709
Benzaldehyde, ≥98%, FG, FCC
C7H6O
W212717
Benzaldehyde, natural, FCC, FG
C7H6O
305197
Benzyl alcohol, anhydrous, 99.8%
C7H8O
ARK2163
Benzyl alcohol, PharmaGrade, USP/NF, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C7H8O
39971
Benzyl alcohol, certified reference material, TraceCERT®
C7H8O
08421
Benzyl alcohol, analytical standard
C7H8O
1061901
Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard
C7H8O
13162
Benzyl alcohol, tested according to Ph.Eur.
C7H8O
PHR1019
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8O
W213705
Benzyl alcohol, ≥99%, FCC, FG
C7H8O
W213713
Benzyl alcohol, natural, ≥98%, FG
C7H8O
Y0000167
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
C7H8O
B6630
Benzyl benzoate, ReagentPlus®, ≥99.0%
C14H12O2
B9550
Benzyl benzoate, meets USP testing specifications
C14H12O2
1062008
Benzyl benzoate, United States Pharmacopeia (USP) Reference Standard
C14H12O2
68183
Benzyl benzoate, analytical standard
C14H12O2
12392
Benzyl benzoate, tested according to Ph.Eur.
C14H12O2
W213802
Benzyl benzoate, ≥99%, FCC, FG
C14H12O2