EMAIL THIS PAGE TO A FRIEND
Bioscience, Biotechnology, and Biochemistry
Diversity in mRNA expression of the serine-type carboxypeptidase ocpG in Aspergillus oryzae through intron retention.
PMID 25130734