EMAIL THIS PAGE TO A FRIEND
Hepatology (Baltimore, Md.)
Bridging the gap between metabolic liver processes and functional tissue structure by integrated spatiotemporal modeling applied to hepatic ammonia detoxification.
PMID 25130974