EMAIL THIS PAGE TO A FRIEND
FEBS letters
Paired related homeobox protein-like 1 (Prrxl1) controls its own expression by a transcriptional autorepression mechanism.
PMID 25131932
Abstract
The homeodomain factor paired related homeobox protein-like 1 (Prrxl1) is crucial for proper assembly of dorsal root ganglia (DRG)-dorsal spinal cord (SC) pain-sensing circuit. By performing chromatin immunoprecipitation with either embryonic DRG or dorsal SC, we identified two evolutionarily conserved regions (i.e. proximal promoter and intron 4) of Prrxl1 locus that show tissue-specific binding of Prrxl1. Transcriptional assays confirm the identified regions can mediate repression by Prrxl1, while gain-of-function studies in Prrxl1 expressing ND7/23 cells indicate Prrxl1 can down-regulate its own expression. Altogether, our results suggest that Prrxl1 uses distinct regulatory regions to repress its own expression in DRG and dorsal SC.
Related Materials