EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Prospective study of surgical treatment of acromegaly: effects on ghrelin, weight, adiposity, and markers of CV risk.
PMID 25137427
Abstract
Although epidemiological studies have found that GH and IGF-1 normalization reduce the excess mortality of active acromegaly to expected rates, cross-sectional data report some cardiovascular (CV) risk markers to be less favorable in remission than active acromegaly. The objective of the study was to test the hypothesis that remission of acromegaly after surgical therapy increases weight and adiposity and some CV risk markers and these changes are paralleled by a rise in ghrelin. Forty-two adults with untreated, active acromegaly were studied prospectively. Changes in outcome measures from before to after surgery were assessed in 26 subjects achieving remission (normal IGF-1) and 16 with persistent active acromegaly (elevated IGF-1) after surgery. The study was conducted at tertiary referral centers for pituitary tumors. Endocrine, metabolic, and CV risk parameters, anthropometrics, and body composition by dual-energy X-ray absorptiometry were measured. Remission increased total ghrelin, body weight, waist circumference, C-reactive protein, homocysteine, high-density lipoprotein, and leptin and reduced systolic blood pressure, homeostasis model assessment score, triglycerides, and lipoprotein (a) by 6 months and for 32 ± 4 months after surgery. The ghrelin rise correlated with the fall in the levels of GH, IGF-1, and insulin and insulin resistance. Weight, waist circumference, and ghrelin did not increase significantly in the persistent active acromegaly group. Total body fat, trunk fat, and perentage total body fat increased by 1 year after surgery in 15 remission subjects: the increase in body fat correlated with the rise in total ghrelin. Although most markers of CV risk improve with acromegaly remission after surgery, some markers and adiposity increase and are paralleled by a rise in total ghrelin, suggesting that these changes may be related. Understanding the mechanisms and long-term implications of the changes that accompany treatment of acromegaly is important to optimizing management because some aspects of the postoperative profile associate with the increased metabolic and CV risk in other populations.