EMAIL THIS PAGE TO A FRIEND
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery
In vivo over-expression of KGF mimic human middle ear cholesteatoma.
PMID 25138153
Abstract
We reported previously that keratinocyte growth factor (KGF), a mesenchymal cell-derived paracrine growth factor, plays an important role in middle ear cholesteatoma formation, which is characterized by marked proliferation of epithelial cells. Here, we investigated whether KGF, the main factor that induces cholesteatoma, overexpression in vivo results in the formation of cholesteatoma. Flag-hKGF cDNA driven by CMV14 promoter was transfected through electroporation into the external auditory canal (EAC) of rats once (short-term model) or five times on every fourth day (long-term model). Ears transfected with empty vector were used as controls. Successful transfection of plasmids into epithelial and stromal cells was confirmed by Flag immunohistochemistry. In the short-term model, the intensity of KGF protein was the strongest in hKGF transfected ear at day 4. KGF expression induced epithelial cell proliferation, reaching a peak level at day 4 and then decreased later, while in the long-term model, KGF expression in the EAC led to middle ear cholesteatoma formation. In conclusion, we described here a new experimental model of human middle ear cholesteatoma, and demonstrated that KGF and KGF receptor paracrine action play an essential role in middle ear cholesteatoma formation in an in vivo model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
OGS1142
PSF-CMV-PURO-COOH-3C-FLAG® - C-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1248
PSF-CMV-PURO-COOH-3C-FLAG®-6HIS - C-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS3407
PSF-CMV-PURO-COOH-EKT-FLAG® - C-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1264
PSF-CMV-PURO-COOH-EKT-FLAG®-6HIS - C-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS3423
PSF-CMV-PURO-COOH-FLAG® - C-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1064
PSF-CMV-PURO-COOH-FXA-FLAG® - C-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1184
PSF-CMV-PURO-COOH-FXA-FLAG®-6HIS - C-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1124
PSF-CMV-PURO-COOH-TEV-FLAG® - C-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1232
PSF-CMV-PURO-COOH-TEV-FLAG®-6HIS - C-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1084
PSF-CMV-PURO-COOH-THR-FLAG® - C-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1200
PSF-CMV-PURO-COOH-THR-FLAG®-6HIS - C-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1247
PSF-CMV-PURO-NH2-6HIS-FLAG®-3C - N-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1263
PSF-CMV-PURO-NH2-6HIS-FLAG®-EKT - N-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1183
PSF-CMV-PURO-NH2-6HIS-FLAG®-FXA - N-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1231
PSF-CMV-PURO-NH2-6HIS-FLAG®-TEV - N-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1199
PSF-CMV-PURO-NH2-6HIS-FLAG®-THR - N-TERMINAL 6 HIS AND FLAG® DUAL TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1493
PSF-CMV-PURO-NH2-BM40-FLAG® - BM40 SECRETION AND FLAG® TAG PLASMID, plasmid vector for molecular cloning
OGS3213
PSF-CMV-PURO-NH2-FLAG® - N-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1141
PSF-CMV-PURO-NH2-FLAG®-3C - N-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS3198
PSF-CMV-PURO-NH2-FLAG®-EKT - N-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1063
PSF-CMV-PURO-NH2-FLAG®-FXA - N-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1123
PSF-CMV-PURO-NH2-FLAG®-TEV - N-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1083
PSF-CMV-PURO-NH2-FLAG®-THR - N-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS1435
PSF-CMV-PURO-NH2-INSULINSP-FLAG® - INSULIN SECRETION AND FLAG® TAG PLASMID, plasmid vector for molecular cloning
OGS3179
PSF-OXB20-COOH-FLAG® - C-TERMINAL FLAG® TAG BACTERIAL PLASMID, plasmid vector for molecular cloning
OGS3317
PSF-OXB20-NH2-FLAG® - N-TERMINAL FLAG® TAG MAMMALIAN PLASMID, plasmid vector for molecular cloning
OGS3098
PSF-OXB20-NH2-OMPA-FLAG® - OMPA SECRETION AND FLAG® TAG PLASMID, plasmid vector for molecular cloning
OGS3156
PSF-OXB20-NH2-PELB-FLAG® - PELB SECRETION AND FLAG® TAG PLASMID, plasmid vector for molecular cloning
OGS3458
PSF-TEF1-COOH-FLAG® - C-TERMINAL FLAG® TAG YEAST PLASMID, plasmid vector for molecular cloning
OGS1874
PSF-TEF1-NH2-A-FACTORFL-FLAG® - ALPHA FACTOR SECRETION AND FLAG® TAG PLASMID, plasmid vector for molecular cloning