EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Hydroxypropyl-β-cyclodextrin grafted polyethyleneimine used as transdermal penetration enhancer of diclofenac sodium.
PMID 25138253
Abstract
The objective of this investigation was to develop a novel cationic polymer, hydroxypropyl-β-cyclodextrin grafted polyethyleneimine (HP-β-CD-PEI1800), as a penetration enhancer, and evaluate its viability on improving transdermal delivery of diclofenac sodium. In this study, HP-β-CD-PEI1800 was characterized by (1)H NMR and DSC methods, respectively. The hydrophilic drug diclofenac sodium was chosen as model drug, and the transdermal permeation enhancement of HP-β-CD-PEI1800 was estimated in vitro by using Franz diffusion cells fitted with mouse dorsal skins, the in vivo kinetics of diclofenac sodium was analyzed by high-performance liquid chromatography (HPLC). The cumulative drug content deposited in epidermis and dermis was measured at the pre-determined time point of 3, 6, and 9h, and the permeation profile was significantly higher than that of the control groups. In addition, the cytotoxicity and skin irritation of enhancer was evaluated by MTT assay and histological examination, respectively, and the results indicated that the polymer we prepared were non-toxic and non-irritant after exposure to skins. All the results suggested that HP-β-CD-PEI1800 could be a safe and efficient penetration enhancer of diclofenac sodium.