EMAIL THIS PAGE TO A FRIEND
Tissue Engineering: Part A
Differential effect of water-soluble chitin on collagen synthesis of human bone marrow stem cells and human periodontal ligament stem cells.
PMID 25138561