EMAIL THIS PAGE TO A FRIEND
Molecular Human Reproduction
The effect of conditioned medium derived from human placental multipotent mesenchymal stromal cells on neutrophils: possible implications for placental infection.
PMID 25140001