EMAIL THIS PAGE TO A FRIEND
Evidence-Based Complementary and Alternative Medicine : ECAM
A Metabonomics Profiling Study on Phlegm Syndrome and Blood-Stasis Syndrome in Coronary Heart Disease Patients Using Liquid Chromatography/Quadrupole Time-of-Flight Mass Spectrometry.
PMID 25140185