EMAIL THIS PAGE TO A FRIEND
Journal of Neurotrauma
Neurological recovery and antioxidant effects of curcumin for spinal cord injury in the rat: a network meta-analysis and systematic review.
PMID 25141070