EMAIL THIS PAGE TO A FRIEND
Development (Cambridge, England)
SirT1 is required in the male germ cell for differentiation and fecundity in mice.
PMID 25142464
Abstract
Sirtuins are NAD(+)-dependent deacylases that regulate numerous biological processes in response to the environment. SirT1 is the mammalian ortholog of yeast Sir2, and is involved in many metabolic pathways in somatic tissues. Whole body deletion of SirT1 alters reproductive function in oocytes and the testes, in part caused by defects in central neuro-endocrine control. To study the function of SirT1 specifically in the male germ line, we deleted this sirtuin in male germ cells and found that mutant mice had smaller testes, a delay in differentiation of pre-meiotic germ cells, decreased spermatozoa number, an increased proportion of abnormal spermatozoa and reduced fertility. At the molecular level, mutants do not have the characteristic increase in acetylation of histone H4 at residues K5, K8 and K12 during spermiogenesis and demonstrate corresponding defects in the histone to protamine transition. Our findings thus reveal a germ cell-autonomous role of SirT1 in spermatogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100019
Anti-ACAA2 antibody produced in rabbit, affinity isolated antibody
HPA042303
Anti-ACAA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105133
Anti-CD5, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA007406
Anti-IL1RL1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101150
Anti-IL1RL1 antibody produced in rabbit, affinity isolated antibody
HPA007917
Anti-IL1RL1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105530
Anti-SLC25A4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0010449M1
Monoclonal Anti-ACAA2 antibody produced in mouse, clone 5C4, purified immunoglobulin, buffered aqueous solution
SAB1404828
Monoclonal Anti-ACAA2 antibody produced in mouse, clone 2F7, purified immunoglobulin, buffered aqueous solution
SAB4100025
Monoclonal Anti-CD5 antibody produced in mouse, culture supernatant
SAB4700066 Monoclonal Anti-CD5 antibody produced in mouse, clone CRIS1, purified immunoglobulin, buffered aqueous solution
SAB4700071 Monoclonal Anti-CD5 antibody produced in mouse, clone MEM-32, purified immunoglobulin, buffered aqueous solution
SAB4700730 Monoclonal Anti-CD5 antibody produced in mouse, clone L17F12, purified immunoglobulin, buffered aqueous solution