EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Cell Physiology
A highly potent agonist to protease-activated receptor-2 reveals apical activation of the airway epithelium resulting in Ca2+-regulated ion conductance.
PMID 25143347