EMAIL THIS PAGE TO A FRIEND
International journal of nanomedicine
Role of surface modification in zinc oxide nanoparticles and its toxicity assessment toward human dermal fibroblast cells.
PMID 25143723