EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Positron emission tomography in CNS drug discovery and drug monitoring.
PMID 25144329
Abstract
Molecular imaging methods such as positron emission tomography (PET) are increasingly involved in the development of new drugs. Using radioactive tracers as imaging probes, PET allows the determination of the pharmacokinetic and pharmacodynamic properties of a drug candidate, via recording target engagement, the pattern of distribution, and metabolism. Because of the noninvasive nature and quantitative end point obtainable by molecular imaging, it seems inherently suited for the examination of a pharmaceutical's behavior in the brain. Molecular imaging, most especially PET, can therefore be a valuable tool in CNS drug research. In this Perspective, we present the basic principles of PET, the importance of appropriate tracer selection, the impact of improved radiopharmaceutical chemistry in radiotracer development, and the different roles that PET can fulfill in CNS drug research.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7752
Serotonin creatinine sulfate monohydrate, powder
C14H19N5O2 · H2O4S · H2O
H4511
Serotonin hydrogen maleate, powder
C10H12N2O · C4H4O4