EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
A review of methods for the analysis of orphan and difficult pesticides: glyphosate, glufosinate, quaternary ammonium and phenoxy acid herbicides, and dithiocarbamate and phthalimide fungicides.
PMID 25145125
Abstract
This article reviews the chromatography/MS methodologies for analysis of pesticide residues of orphan and difficult chemical classes in a variety of sample matrixes including water, urine, blood, and food. The review focuses on pesticide classes that are not commonly included in multiresidue analysis methods such as highly polar or ionic herbicides including glyphosate, glufosinate, quaternary ammonium, and phenoxy acid herbicides, and some of their major degradation or metabolite products. In addition, dithiocarbamate and phthalimide fungicides, which are thermally unstable and have stability issues in some solvents or sample matrixes, are also examined due to their special needs in residue analysis.