EMAIL THIS PAGE TO A FRIEND
BJOG : an international journal of obstetrics and gynaecology
There is still insufficient data to recommend screening twin pregnancies with transvaginal ultrasound cervical length, but this might change soon!
PMID 25145404