EMAIL THIS PAGE TO A FRIEND
Tissue Engineering: Part A
Physical and biological characterization of ferromagnetic fiber networks: effect of fibrin deposition on short-term in vitro responses of human osteoblasts.
PMID 25145466