EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Determination of solubility parameters of ionic liquids and ionic liquid/solvent mixtures from intrinsic viscosity.
PMID 25145759
Abstract
The total and partial solubility parameters (dispersion, polar and hydrogen-bonding solubility parameters) of ten ionic liquids were determined. Intrinsic viscosity approaches were used that encompassed a one-dimensional method (1D-Method), and two different three-dimensional methods (3D-Method1 and 3D-Method2). The effect of solvent type, the dimethylacetamide (DMA) fraction in the ionic liquid, and dissolution temperature on solubility parameters were also investigated. For all types of effect, both the 1D-Method and 3D-Method2 present the same trend in the total solubility parameter. The partial solubility parameters are influenced by the cation and anion of the ionic liquid. Considering the effect on partial solubility parameters of the solvent type in the ionic liquid, it was observed that in both 3D methods, the dispersion and polar parameters of a 1-ethyl-3-methylimidazolium acetate/solvent (60:40 vol %) mixture tend to increase as the total solubility parameter of the solvent increases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7906
1-Butanol, for molecular biology, ≥99%
C4H10O
19422
1-Butanol, analytical standard
C4H10O
281549
1-Butanol, anhydrous, 99.8%
C4H10O
270679
1-Butanol, for HPLC, 99.8%
C4H10O
442665
1-Butanol, analytical standard
C4H10O
03-4585
1-Butanol, for HPLC
C4H10O
PHR1209
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
C4H10O
04129
1-Butyl-3-methylimidazolium chloride, ≥99.0% (HPLC)
C8H15ClN2
94128
1-Butyl-3-methylimidazolium chloride, ≥98.0% (HPLC)
C8H15ClN2
38899
1-Butyl-3-methylimidazolium chloride, ≥98.0% (HPLC)
C8H15ClN2
689483
1-Ethyl-3-methylimidazolium acetate, 97%
C8H14N2O2
51053
1-Ethyl-3-methylimidazolium acetate, ≥95.0% (HPLC)
C8H14N2O2
671541
1-Ethyl-3-methylimidazolium diethyl phosphate, ≥98.0% (HPLC)
C10H21N2O4P
713392
1-Ethyl-3-methylimidazolium diethyl phosphate, ≥98.0%
C10H21N2O4P
279544
1-Propanol, anhydrous, 99.7%
C3H8O
96566
1-Propanol, analytical standard
C3H8O
W292818
1-Propanol, ≥99%, FG
C3H8O
W292826
1-Propanol, natural, ≥98%, FG
C3H8O
24-6080
1-Propanol, SAJ first grade, ≥99.0%
C3H8O
24-6070
1-Propanol, JIS special grade
C3H8O
1570406
1-Propanol, United States Pharmacopeia (USP) Reference Standard
C3H8O
PHR1208
1-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O
294810
2-Butanol, anhydrous, 99.5%
C4H10O
96870
2-Butanol, analytical standard
C4H10O
PHR1563
2-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
C4H10O
19440
2-Butanol, puriss. p.a., Reag. Ph. Eur., ≥99.5% (GC)
C4H10O
B85919
2-Butanol, ReagentPlus®, ≥99%
C4H10O
240338
2-Pyrrolidinone, ≥99%
C4H7NO
P74370
2-Pyrrolidinone, 99%
C4H7NO
83300
2-Pyrrolidinone, purum, ≥98.0% (GC)
C4H7NO