EMAIL THIS PAGE TO A FRIEND
Journal of Neuroscience Research
The selective noradrenergic reuptake inhibitor reboxetine restores spatial learning deficits, biochemical changes, and hippocampal synaptic plasticity in an animal model of depression.
PMID 25146499