EMAIL THIS PAGE TO A FRIEND
Clinical and experimental rheumatology
Blockade of macrophage autophagy ameliorates activated lymphocytes-derived DNA induced murine lupus possibly via inhibition of proinflammatory cytokine production.
PMID 25151985
Abstract
Systemic lupus erythematosus (SLE) is a typical inflammatory autoimmune disease for its unknown pathogenesis and potential fatality. It has been reported that autophagy has a crosstalk with autoimmunity, but its impact on the pathogenesis of SLE remains unclear. Here, we investigated the role of autophagy in inflammatory response of macrophages under SLE conditions. First, we detected the expression of autophagy-related genes (Atg5, Atg12 and Beclin 1) in the macrophages derived from activated lymphocytes-derived DNA (ALD-DNA) induced murine lupus as well as in the PBMC from SLE patients. And then through adoptive transfer of Beclin 1 knockdown macrophages, we further investigated the potential effect of macrophage autophagy on the SLE-associated inflammatory response and disease severity by evaluating serum anti-dsDNA antibodies and proteinuria levels, immune complex deposition as well as renal pathological changes. We found that autophagy related genes were significantly upregulated in the splenic and renal macrophages of lupus mice and in the PBMC of SLE patients. Adoptive transfer of Beclin 1 knockdown macrophages could significantly decrease the anti-dsDNA antibodies and proteinuria levels, robustly reduce renal immune complex deposition and remit glomerulonephritis, indicating the amelioration of murine lupus. This protective effect was associated with the obviously decreased production of proinflammatory cytokines IL-6 and TNF-α. Our results suggested that aberrant activated autophagy in macrophages contributed to the pathogenesis of murine lupus possibly via promoting the production of proinflammatory cytokines TNF-α and IL-6, and inhibition of autophagy might represent a novel regulation strategy for excessive activation of proinflammatory macrophages and a new therapeutic regime for SLE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4434
Dichloromethylenediphosphonic acid disodium salt
CH2Cl2Na2O6P2