EMAIL THIS PAGE TO A FRIEND
International Journal of Obesity
Grape seed procyanidin supplementation to rats fed a high-fat diet during pregnancy and lactation increases the body fat content and modulates the inflammatory response and the adipose tissue metabolism of the male offspring in youth.
PMID 25152240