EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Expression of cGMP-dependent protein kinase type I in mature white adipocytes.
PMID 25152402
Abstract
The presence of cGMP-dependent protein kinase I (cGKI) in murine adipocytes has been questioned, although cGKI was implicated in the thermogenic program of fat cells (FCs) and to exert anti-hypertrophic/-inflammatory effects in white adipose tissue. Herein, cGKI was detected in adipocytes from control mice, whereas FCs from global cGKI knockouts (cGKI(-/-)) and cGKIα rescue (αRM) mice remained cGKI-negative. cGKI mutants exhibit decreased adipocyte size, plasma leptin levels and reduced body-weights as compared to litter-matched controls. Low abundance of adiponectin in WAT and plasma of αRM animals together with previously confirmed high IL-6 levels indicate a low-grade inflammation. However, αRMs were protected from streptozotocin-induced hyperglycemia. Our results suggest that cGMP/cGKI affects both glucose and FC homeostasis in more complex mode than previously thought.