EMAIL THIS PAGE TO A FRIEND
Seizure : the Journal of the British Epilepsy Association
Correlation of preoperative seizures with clinicopathological factors and prognosis in anaplastic gliomas: a report of 198 patients from China.
PMID 25153654