EMAIL THIS PAGE TO A FRIEND
Antimicrobial Agents and Chemotherapy
Maternal intravenous administration of azithromycin results in significant fetal uptake in a sheep model of second trimester pregnancy.
PMID 25155606