EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Position and length of fatty acids strongly affect receptor selectivity pattern of human pancreatic polypeptide analogues.
PMID 25156249
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90968
1,2-Ethanedithiol, analytical standard
C2H6S2
02390
1,2-Ethanedithiol, ≥98.0% (GC)
C2H6S2
398020
1,2-Ethanedithiol, technical grade, ≥90%
C2H6S2
T28525
4-Methylbenzenethiol, 98%
C7H8S
92337
Bromotrimethylsilane, purum, ≥97.0% (AT)
C3H9BrSi
194409
Bromotrimethylsilane, 97%
C3H9BrSi
445460
HATU, 97%
C10H15F6N6OP
W387304
Methyl phenyl sulfide, ≥99%
C7H8S
03440
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
C8H19N
03439
N-Ethyldiisopropylamine solution, for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
C8H19N
43051
Palmitic acid, certified reference material, TraceCERT®
C16H32O2
1492007
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
C16H32O2
76119
Palmitic acid, analytical standard
C16H32O2
76122
Palmitic acid, purum, ≥98.0% (GC)
C16H32O2
PHR1120
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C16H32O2
P0090000
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
C16H32O2
411027
Piperidine, ≥99.5%, purified by redistillation
C5H11N
104094
Piperidine, ReagentPlus®, 99%
C5H11N
76046
Piperidine, analytical standard
C5H11N
80642
Piperidine, purum, ≥96.0% (GC/T)
C5H11N
W290807
Piperidine, ≥99%, FCC
C5H11N
80645
Piperidine solution, for peptide synthesis, 20% in DMF
C5H11N
88470
Thioanisole, ReagentPlus®, ≥99%
C7H8S
T28002
Thioanisole, ReagentPlus®, ≥99%
C7H8S
92358
Thioanisole, analytical standard
C7H8S