EMAIL THIS PAGE TO A FRIEND
Bioorganic & Medicinal Chemistry
Shogaol-huprine hybrids: dual antioxidant and anticholinesterase agents with ?-amyloid and tau anti-aggregating properties.
PMID 25156301
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8130
5,5′-Dithiobis(2-nitrobenzoic acid), BioReagent, ≥98%, suitable for determination of sulfhydryl groups
C14H8N2O8S2
D218200
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
C14H8N2O8S2
A5751
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
C7H16INOS
01480
Acetylthiocholine iodide, ≥99.0% (AT)
C7H16INOS
G7384
Gallic acid, 97.5-102.5% (titration)
C7H6O5
91215
Gallic acid, certified reference material, TraceCERT®
C7H6O5
529664
Manganese(IV) oxide, ≥99.99% trace metals basis
MnO2
203750
Manganese(IV) oxide, 99.99% trace metals basis
MnO2
243442
Manganese(IV) oxide, ReagentPlus®, ≥99%
MnO2
310700
Manganese(IV) oxide, 10 μm, reagent grade, ≥90%
MnO2
217646
Manganese(IV) oxide, activated, ~85%, <10 μm
MnO2
775711
Manganese(IV) oxide, nanorod, diam. × L 5-30 nm × 80-100 nm, 99% trace metals basis (excludes potassium content)
MnO2
B3253
S-Butyrylthiocholine iodide, ≥98%
C9H20INOS
20820
S-Butyrylthiocholine iodide, puriss., ≥99.0% (AT)
C9H20INOS
791768
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
71363
Sodium carbonate, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
CNa2O3
72784
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
CNa2O3
56169
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
CNa2O3
T1892 Thioflavine S, practical grade