EMAIL THIS PAGE TO A FRIEND
Talanta
Ethanol as an alternative to formaldehyde for the enhancement of manganese(IV) chemiluminescence detection.
PMID 25159402
Abstract
Previous applications of manganese(IV) as a chemiluminescence reagent have required the use of formaldehyde to enhance the emission intensity to analytically useful levels. However, this known human carcinogen (by inhalation) is not ideal for routine application. A wide range of alternative enhancers have been examined but to date none have been found to provide the dramatic increase in chemiluminescence intensities obtained using formaldehyde. Herein, we demonstrate that ethanol offers a simple, safe and inexpensive alternative to the use of formaldehyde for manganese(IV) chemiluminescence detection, without compromising signal intensity or sensitivity. For example, chemiluminescence signals for opiate alkaloids using 50-100% ethanol were 0.8-1.6-fold those using 2M formaldehyde. This innocuous alternative enhancer is shown to be a particularly effective for the direct detection of thiols and disulfides by manganese(IV) chemiluminescence, which we have applied to a simple HPLC procedure to determine a series of biomarkers of oxidative stress.