EMAIL THIS PAGE TO A FRIEND
Talanta
Enantioselective HPLC-DAD method for the determination of etodolac enantiomers in tablets, human plasma and application to comparative pharmacokinetic study of both enantiomers after a single oral dose to twelve healthy volunteers
PMID 25159440
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K08971227 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 50 mm × 2.1 mm
K08971223 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 150 mm × 2.1 mm
K08971228 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 50 mm × 2.1 mm
K08971224 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 150 mm × 2.1 mm
K08670372 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 50 mm × 4.6 mm
K08670370 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 150 mm × 4.6 mm
K08670617 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 50 mm × 4.6 mm
K08670373 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 150 mm × 4.6 mm
K08670374 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 4.6 mm
K08670614 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 10  mm
K08670615 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 21.2 mm
K08670616 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 30 mm
K08670612 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 150 mm × 4.6 mm
K08670613 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 4.6 mm
K08670609 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 10 mm
K08670610 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 21.2 mm
K08670611 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 30 mm
K08670502 Kromasil® CelluCoat® Chiral Reversed Phase HPLC Column, 3 μm particle size, L × I.D. 50 mm × 4.6 mm
K08670503 Kromasil® CelluCoat® Chiral Reversed Phase HPLC Column, 3 μm particle size, L × I.D. 150 mm × 4.6  mm