EMAIL THIS PAGE TO A FRIEND
British Journal of Pharmacology
Effects of caffeine on circadian phase, amplitude and period evaluated in cells in vitro and peripheral organs in vivo in PER2::LUCIFERASE mice.
PMID 25160990