EMAIL THIS PAGE TO A FRIEND
Molecular and Cellular Endocrinology
Melatonin and its metabolites accumulate in the human epidermis in vivo and inhibit proliferation and tyrosinase activity in epidermal melanocytes in vitro.
PMID 25168391