EMAIL THIS PAGE TO A FRIEND
International Journal of Pharmaceutics
Ligand density at the surface of a nanoparticle and different uptake mechanism: two important factors for successful siRNA delivery to liver endothelial cells.
PMID 25169077